missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serine racemase Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06588CL1
This item is not returnable.
View return policy
Description
Serine racemase Polyclonal antibody specifically detects Serine racemase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Serine racemase | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| D-serine ammonia-lyase, D-serine dehydratase, EC 4.3.1.17, EC 4.3.1.18, EC 5.1.1.18, ILV1, ISO1, L-serine ammonia-lyase, L-serine dehydratase, serine racemase | |
| Produced in rabbits immunized with E. coli-derived Human Serine racemase fragment. | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 63826 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction