missing translation for 'onlineSavingsMsg'
Learn More

Serine Palmitoyltransferase 2 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18419087 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419087 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419087 Supplier Novus Biologicals Supplier No. NBP176573C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Serine Palmitoyltransferase 2 Polyclonal antibody specifically detects Serine Palmitoyltransferase 2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serine Palmitoyltransferase 2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias EC 2.3.1, EC 2.3.1.50, KIAA0526HSN1C, LCB 2, LCB2a, LCB2NSAN1C, Long chain base biosynthesis protein 2, Long chain base biosynthesis protein 2a, serine palmitoyltransferase 2, serine palmitoyltransferase, long chain base subunit 2, serine palmitoyltransferase, subunit II, Serine-palmitoyl-CoA transferase 2, SPT 2, SPT2
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of human Serine Palmitoyltransferase 2. The immunogen is located within the last 50 amino acids of Serine Palmitoyltransferase 2. Amino Acid Sequence: LDRPFDETTYEETED
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9517
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.