missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serine Dehydratase Antibody (OTI3D3), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74074MFV610
This item is not returnable.
View return policy
Description
Serine Dehydratase Monoclonal antibody specifically detects Serine Dehydratase in Human samples. It is validated for Flow Cytometry
Specifications
| Serine Dehydratase | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 10993 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI3D3 | |
| Flow Cytometry | |
| EC 4.3.1.17, EC 4.3.1.19, L-serine ammonia-lyase, L-serine deaminase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, SDHL-serine dehydratase, serine dehydratase, TDH | |
| Full length human recombinant protein of human SDS(NP_006834) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction