missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) Series S Sensor Chip NA

Product Code. 17049203 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Application):
Sensor Chip for High-Affinity Capture of Biotinylated Molecules
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
17049203 Sensor Chip for High-Affinity Capture of Biotinylated Molecules Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17049203 Supplier Cytiva (Formerly GE Healthcare Life Sciences) Supplier No. 29407997
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For capture of biotinylated molecules with analysis of ligand-analyte binding in low MW applications

  • Reduced nonspecific binding: an alternative to test when nonspecific binding effects are experienced with Sensor Chip SA
  • Noncovalent: convenient alternative for ligands sensitive to covalent immobilization
  • Ready-to-use: pre-immobilized NeutrAvidin™ sensor chip eliminates need for preparation of coupling reagents and optimization of coupling conditions
  • Versatile: immobilize biotinylated peptides, proteins, or nucleic acids for oriented capture
  • Robust: reproducible high-affinity ligand capture and robust capacity for more than 200 cycles
  • Flexible: supports researchers with preference for NeutrAvidin™ ligand
TRUSTED_SUSTAINABILITY

Specifications

Kit Contents 1 Sensor Chip
Content And Storage 2°C to 8°C in unopened pouches
For Use With (Equipment) Biacore 8 K+, Biacore 8 K, Biacore T200, Biacore S200, Biacore 4000 SPR Systems
Surface Functionality Carboxymethylated dextran pre-immobilized with NeutrAvidin™
For Use With (Application) Sensor Chip for High-Affinity Capture of Biotinylated Molecules
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.