Learn More
3M™ Series G3000 Ventilated Ratchet Hard Hat
Series G3000 Ventilated Ratchet Hard Hat allows falling objects to slide off easily and not dislodge the helmet from its position.
Brand:  3M™ 7000009701_CLEAN
Product Code. 15719647
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • A hard hat designed for use in harsh environments, with excellent ventilation and a good field of vision.
  • 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV- radiation and its time to replace it.
  • Softly rounded design, which helps to avoid helmet parts snagging on branches, etc.
  • Ventilation holes distributed over a larger area offer maximum through-flow of air increasing wearer comfort.
  • Head harness can be rotated 180° for better visibility when working in confined spaces.
  • Short peak gives improved upwards visibility.