missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Series CBF CO2 Incubators with Hot Air Sterilization and Humidity Regulation
GREENER_CHOICE

Product Code. 16377956 Shop All BINDER Products
Click to view available options
Capacity (Metric):
170 L
Model:
CB170-230V-OF
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16377956

Brand: BINDER™ 90400134

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The BINDER CBF is the high-end CO2 incubator with humidity control that is suitable for all sensitive incubation applications in GMP environments. It ensures consistent results including in cell-based assays with multiwell plates.

This BINDER incubator is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.
  • Contamination-safe humidification system without water pan
  • Tried-and-tested anti-contamination concept with 180 °C hot air sterilization
  • Hot-air sterilizable CO2 sensor with single-beam infrared technology
  • Seamless inner chamber made of stainless steel with flanges as shelf support system
  • Fanless interior with Venturi CO2 gas mixing nozzle
  • Intuitive touchscreen controller
  • Internal data logger, measured values can be read out in open format via USB
  • Troubleshooting system with visual and acoustic alarms
  • Interfaces: Ethernet, USB, zero-voltage alarm contact
  • Fail-safe CO2 system for protecting the pH of cell cultures

Specifications

Description O2 Control 0.2 to 20% and Cell Therapy Compartmentalization
Capacity (Metric) 170 L
Temperature Range (Metric) Ambient +5° to 60°C
Display TFT Touch
Model CB170-230V-OF
Dimensions (D x W x H) Interior 560 x 505 x 600 mm
Dimensions (L x W x H) 685 x 815 x 870 mm
Weight (Metric) 95 kg
Voltage 230 V
Wattage 1.4 kW
Amperage 5.7 A
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.