missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Series CBF CO2 Incubator, with Hot Air Sterilization and Humidity Regulation
GREENER_CHOICEBIOPHARMA

Product Code. 17353703 Shop All BINDER Products
Click to view available options
Capacity (Metric):
170 L
267 L
Model:
CBF170-230 V
CBF170-230 V-O
CBF260-230 V
CBF260-230 V-O
CBF260-230 V-OZ
Oxygen Control:
None
Yes
Unit Size:
Each
This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Product Code. 17353703

Brand: BINDER™ 96400018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

The CBF is the high-end CO2 incubator with humidity control that is suitable for all sensitive incubation applications in GMP environments. It ensures consistent results including in cell-based assays with multiwell plates.

This BINDER incubator is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.
  • Contamination-safe humidification system without water pan
  • Tried-and-tested anti-contamination concept with 180°C hot air sterilization
  • Hot-air sterilizable CO2 sensor with single-beam infrared technology
  • Seamless inner chamber made of stainless steel with flanges as shelf support system
  • Fanless interior with Venturi CO2 gas mixing nozzle
  • Intuitive touchscreen controller
  • Internal data logger, measured values can be read out in open format via USB
  • Troubleshooting system with visual and acoustic alarms
  • Interfaces: Ethernet, USB, zero-voltage alarm contact
  • Fail-safe CO2 system for protecting the pH of cell cultures

Specifications

Capacity (English) 6 cu. ft.
Capacity (English) Shelf 22 lb.
Capacity (Metric) Shelf 10 kg
Description CBF170-230 V-O, with O2 control 0.2-20%, Series CBF - CO2 incubators with hot air sterilization and humidity regulation, 170 L, 230 V 1 ∼ 50/60 Hz
Capacity (Metric) 170 L
Chamber Material Stainless Steel
CO2 Concentration Range 0 to 20%
CO2 Sensor Technology IR Sensor
Convection Type Natural, Fanless Interior
Holds None
Lighting No Lighting
Material Stainless Steel
No. of Places 1
No. of Shelves 3
O2 Concentration Range 0.2 to 20%
Oxygen Control Yes
Plug Type Nema 5-20P
Relative Humidity 90 to 95%
Temperature Range (Metric) Ambient +6° to 60°C
Type CO2 Incubator
Display TFT touch
Model CBF170-230 V-O
O2 Sensor Technology IR sensor
Sensor Types IR sensor
Dimensions (D x W x H) Interior 560 x 505 x 600 mm
Dimensions (L x W x H) 685 x 815 x 870 mm
Weight (Metric) 95 kg
Voltage 230 V
Wattage 1.4 kW
Amperage 5.7 A
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.