missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERGEF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558116
This item is not returnable.
View return policy
Description
Immunogen sequence: GTEANVWAPK PVQALLSSSG LLVGCGAGHS LALCQLPAHP ALVQDPKVTY LSPDAIEDTE SQKAMDKERN WKERQSETST QSQSDWSRNG Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 56%.
Probable guanine nucleotide exchange factor (GEF), which may be involved in the secretion process.
Specifications
| SERGEF | |
| Polyclonal | |
| Unconjugated | |
| SERGEF | |
| deafness locus associated putative guanine nucleotide exchange factor; deafness locus-associated putative guanine nucleotide exchange factor; DelGEF; Gef; Gnef; Gnefr; guanine nucleotide exchange factor (RCC1 related); guanine nucleotide exchange factor-related protein; secretion regulating guanine nucleotide exchange factor; secretion-regulating guanine nucleotide exchange factor; Sergef | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26297 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UGK8 | |
| SERGEF | |
| Recombinant protein corresponding to Human SERGEF. Recombinant protein control fragment (Product #RP-97325). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction