missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SERF2 Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Specifications
Specifications
| Gene ID (Entrez) | 10169 |
| Species | Human |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-78375. For further blocking peptide related protocol, click here . |
| Gene Alias | 4F5rel, FAM2CFLJ38557,4F5REL, FLJ20431, FLJ37527, Gastric cancer-related protein VRG107, H4F5REL, HsT17089, MGC48826, Protein 4F5-related, small EDRK-rich factor 2 |
| Gene Symbol | SERF2 |
| Quantity | 0.05 mg |
| Research Category | Neuroscience |
| Storage Buffer | Peptide dissolved in dH2O. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction