missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERF1A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £386.00
Specifications
| Antigen | SERF1A |
|---|---|
| Concentration | 1.14 mg/mL |
| Dilution | Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:400 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247387
|
Novus Biologicals
NBP1-78393SS |
0.025 mL |
£169.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18279555
|
Novus Biologicals
NBP1-78393 |
0.1 mL |
£386.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SERF1A Polyclonal specifically detects SERF1A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SERF1A | |
| Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:400 | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Neuroscience | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| FAM2B, h4F5, H4F5small EDRK-rich factor 1,4F5, Protein 4F5, SERF1, SMA modifier 1, small EDRK-rich factor 1A (telomeric), SMAM1FAM2ASERF1B, spinal muscular atrophy-related gene H4F5 | |
| SERF1A | |
| IgG | |
| Affinity Purified |
| 1.14 mg/mL | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| O75920 | |
| 8293 | |
| A synthetic peptide made to an internal portion of the human SERF1A protein (between residues 50-100) [UniProt O75920] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title