missing translation for 'onlineSavingsMsg'
Learn More

SERF1A Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18370551 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370551 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370551 Supplier Novus Biologicals Supplier No. NBP178392AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SERF1A Polyclonal specifically detects SERF1A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERF1A
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FAM2B, h4F5, H4F5small EDRK-rich factor 1,4F5, Protein 4F5, SERF1, SMA modifier 1, small EDRK-rich factor 1A (telomeric), SMAM1FAM2ASERF1B, spinal muscular atrophy-related gene H4F5
Gene Symbols SERF1A
Host Species Rabbit
Immunogen A synthetic peptide made to the N-terminal portion of the human SERF1A protein (between residues 1-60) [UniProt O75920]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8293
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.