missing translation for 'onlineSavingsMsg'
Learn More

SERCA3 ATPase Antibody (1B9), Novus Biologicals™

Product Code. 18356397 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356397 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356397 Supplier Novus Biologicals Supplier No. H00000489M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SERCA3 ATPase Monoclonal antibody specifically detects SERCA3 ATPase in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERCA3 ATPase
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1B9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH35729
Gene Alias adenosine triphosphatase, calcium, ATPase, Ca(2+)-transporting, ubiquitous, ATPase, Ca++ transporting, ubiquitous, Calcium pump 3, EC 3.6.3, EC 3.6.3.8, sarco/endoplasmic reticulum Ca2+ ATPase, sarco/endoplasmic reticulum Ca2+ -ATPase, sarcoplasmic/endoplasmic reticulum calcium ATPase 3, SERCA3calcium-translocating P-type ATPase, SR Ca(2+)-ATPase 3
Host Species Mouse
Immunogen ATP2A3 (AAH35729, 501 a.a. ~ 620 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TRPHPTGQGSKMFVKGAPESVIERCSSVRVGSRTAPLTPTSREQILAKIRDWGSGSDTLRCLALATRDAPPRKEDMELDDCSKFVQYETDLTFVGCVGMLDPPRPEVAACITRCYQAGIR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 489
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.