missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERCA2 ATPase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584558
This item is not returnable.
View return policy
Description
Immunogen sequence: TVEEVLGHFG VNESTGLSLE QVKKLKERWG SNELPA Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
This gene encodes one of the SERCA Ca(2+)-ATPases, which are intracellular pumps located in the sarcoplasmic or endoplasmic reticula of muscle cells. This enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol into the sarcoplasmic reticulum lumen, and is involved in regulation of the contraction/relaxation cycle. Mutations in this gene cause Darier-White disease, also known as keratosis follicularis, an autosomal dominant skin disorder characterized by loss of adhesion between epidermal cells and abnormal keratinization. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| SERCA2 ATPase | |
| Polyclonal | |
| Unconjugated | |
| Atp2a2 | |
| 9530097L16Rik; ATP2; Atp2a2; ATP2B; ATPase Ca++ transporting cardiac muscle slow twitch 2; ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2; ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2; ATPase, Ca++ transporting, cardiac muscle, slow twitch 2; ATPase, Ca++ transporting, slow twitch 2; Ca(2+)-transport ATPase class 3; calcium ATPase; calcium pump 2; calcium-ATPase (EC 3.6.1.3); calcium-transporting ATPase; calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; cardiac Ca2+ ATPase; D5Wsu150e; DAR; DD; endoplasmic reticulum class 1/2 Ca(2+) ATPase; mKIAA4195; putative SERCA isoform; sarco(endo)plasmic reticulum Ca(2+)-dependent ATPase 2; sarco/endoplasmic reticulum Ca2+-ATPase 2; sarcoplasmic reticulum Ca2+-transport ATPase isoform; sarcoplasmic/endoplasmic reticulum calcium ATPase 2; sarcoplasmic/endoplasmic-reticulum Ca(2+) pump gene 2; SERCA; SERCA ATPase; SERCA2; Serca2a; SERCA2B; SercaII; SR Ca(2+)-ATPase 2; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 488 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P16615 | |
| Atp2a2 | |
| Recombinant protein corresponding to Human ATP2A2. Recombinant protein control fragment (Product #RP-101984). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction