missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERCA2 ATPase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£180.00 - £248.00
Specifications
| Antigen | SERCA2 ATPase |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:100-1:400 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682522
|
Novus Biologicals
NBP2-97582-100ul |
100 μg |
£248.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627714
|
Novus Biologicals
NBP2-97582-50ul |
50 μg |
£180.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SERCA2 ATPase Polyclonal antibody specifically detects SERCA2 ATPase in Human, Cynomolgus Monkey samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| SERCA2 ATPase | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS (pH 7.0) | |
| 488 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:100-1:400 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Cynomolgus Monkey | |
| ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2 | |
| Produced in rabbits immunized with E. coli-derived Human SERCA2 ATPase fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title