missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERCA2 ATPase Antibody (IID8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-529CL1
This item is not returnable.
View return policy
Description
SERCA2 ATPase Monoclonal antibody specifically detects SERCA2 ATPase in Amphibian,Bovine,Canine,Guinea Pig,Hamster,Human,Mouse,Porcine,Primate,Rabbit,Rat,Sheep,Virus samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Neutralization,Immunocytochemistry/Immunofluorescence
Specifications
| SERCA2 ATPase | |
| Monoclonal | |
| CoraFluor 1 | |
| ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2 | |
| Purified canine cardiac sarcoplasmic reticulum. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 488 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| IID8 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Amphibian, Bovine, Canine, Guinea Pig, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction