missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERCA2 ATPase Antibody (IID8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB300-529
This item is not returnable.
View return policy
Description
SERCA2 ATPase Monoclonal specifically detects SERCA2 ATPase in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.
Specifications
| SERCA2 ATPase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:5000, Flow Cytometry 1 ug 10^6 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1 ug/ml, Immunohistochemistry-Frozen 1:500, Block/Neutralize | |
| ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 488 | |
| Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Virus | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| IID8 | |
| Unconjugated | |
| O46674 | |
| ATP2A2 | |
| Purified canine cardiac sarcoplasmic reticulum. | |
| 100 μL | |
| Primary | |
| Detects Sarcoplasmic or Endoplasmic Reticulum Calcium 2 (SERCA2) ATPase. This does not discriminate between the two isoforms. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction