missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERCA1 ATPase Recombinant Rabbit Monoclonal Antibody (23GB4340), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552693
This item is not returnable.
View return policy
Description
ATP dependent calcium pumps are responsible, in part, for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium (SERCA) ATPases. The SERCA1 gene is exclusively expressed in type II (fast) skeletal muscle. The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific form expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. The SERCA3 gene is not as well characterized and is found in non-muscle cells.
Specifications
| SERCA1 ATPase | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ATP2A1 | |
| adult Ca2+ ATPase; ATP2A; ATP2A1; ATP2A3; ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1; ATPase, Ca++ transporting, cardiac muscle, fast twitch 1; ATPase, Ca++ transporting, ubiquitous; Ca2+ ATPase; calcium pump 1; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; endoplasmic reticulum class 1/2 Ca(2+) ATPase; neonatal Ca2+ ATPase; sarcoplasmic/endoplasmic reticulum calcium ATPase 1; SERCA; SERCA ATPase; Serca1; SERCA1a; SR Ca(2+)-ATPase 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 487 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 23GB4340 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14983 | |
| ATP2A1 | |
| A synthesized peptide derived from human ATP2A1/SERCA1 (1-54AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction