missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals SERCA1 ATPase Antibody (6F8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00000487-M02
This item is not returnable.
View return policy
Description
SERCA1 ATPase Monoclonal antibody specifically detects SERCA1 ATPase in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| SERCA1 ATPase | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| ATP2A, ATPase, Ca++ transporting, cardiac muscle, fast twitch 1, Calcium pump 1, Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletalmuscle isoform, EC 3.6.3, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, sarcoplasmic/endoplasmic reticulum calcium ATPase 1, SERCA1EC 3.6.3.8, SR Ca(2+)-ATPase 1 | |
| ATP2A1 (NP_775293, 522 a.a. ~ 612 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IDRCNYVRVGTTRVPLTGPVKEKIMAVIKEWGTGRDTLRCLALATRDTPPKREEMVLDDSARFLEYETDLTFVGVVGMLDPPRKEVTGSIQ | |
| 0.1 mg | |
| Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 487 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 6F8 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_775293 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction