missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERBP1, Mouse anti-Human, Clone: SERBP1/3496, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79710-20UG
This item is not returnable.
View return policy
Description
SERBP1 Monoclonal specifically detects SERBP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| SERBP1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Q8NC51 | |
| SERBP1 | |
| Recombinant fragment of human SERBP1 protein (around aa3-139) (exact sequence is proprietary) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| SERBP1/3496 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| CGI-55, CHD3IP, chromodomain helicase DNA binding protein 3 interacting protein, DKFZP564M2423, HABP4L, PAI-1 mRNA binding protein, PAI1 RNA-binding protein 1, PAI-RBP1DKFZp564M2423, PAIRBP1FLJ90489, plasminogen activator inhibitor 1 RNA binding protein, plasminogen activator inhibitor 1 RNA-binding protein, SERPINE1 mRNA binding protein 1, SERPINE1 mRNA-binding protein 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 26135 | |
| Store at 4°C. | |
| IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction