missing translation for 'onlineSavingsMsg'
Learn More

Seratrodast, TRC

Product Code. 30420616
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420616 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420616 Supplier Toronto Research Chemicals Supplier No. TRCS2630005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Seratrodast
CAS 112665-43-7
Molecular Formula C22 H26 O4
Quantity 5 mg
Synonym Benzeneheptanoic acid','ζ-(2,4,5-trimethyl-3,6-dioxo-1,4-cyclohexadien-1-yl)-, Benzeneheptanoic acid','ζ-(2,4,5-trimethyl-3,6-dioxo-1,4-cyclohexadien-1-yl)-','(±)-, ζ-(2,4,5-Trimethyl-3,6-dioxo-1,4-cyclohexadien-1-yl)benzeneheptanoic acid, (±)-2,4,5-Trimethyl-3,6-dioxo-ζ-phenyl-1,4-cyclohexadiene-1-heptanoic acid, 7-(3,5,6-Trimethyl-1,4-benzoquinon-2-yl)-7-phenylheptanoic acid, A 73001, AA 2414, ABT 001, Abbott 73001, Bronica, Seratodrast, Seratrodast
InChI Formula InChI=1S/C22H26O4/c1-14-15(2)22(26)20(16(3)21(14)25)18(17-10-6-4-7-11-17)12-8-5-9-13-19(23)24/h4,6-7,10-11,18H,5,8-9,12-13H2,1-3H3,(H,23,24)
SMILES CC1=C(C)C(=O)C(=C(C)C1=O)C(CCCCCC(=O)O)c2ccccc2
IUPAC Name 7-phenyl-7-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)heptanoic acid
Molecular Weight (g/mol) 354.44
Formula Weight 354.1831
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.