missing translation for 'onlineSavingsMsg'
Learn More

Seratrodast, MedChemExpress

Product Code. 16484477
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484477 50 mg 50mg
16428608 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16484477 Supplier Medchem Express Supplier No. HYB0774

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Seratrodast(AA 2414) is an anti-asthmatic agent and a potent and selective thromboxane A2 receptor (TP) antagonist.

Seratrodast(AA 2414) is an anti-asthmatic agent and a potent and selective thromboxane A2 receptor (TP) antagonist.

Specifications

Chemical Name or Material Seratrodast
Physical Form Solid
CAS 112665-43-7
Color Orange
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H26O4
For Use With (Application) COVID-19-immunoregulation
Synonym AA 2414
Solubility Information DMSO : ≥ 100 mg/mL (282.14 mM)
SMILES CC(C1=O)=C(C(C(C)=C1C)=O)C(C2=CC=CC=C2)CCCCCC(O)=O
Molecular Weight (g/mol) 354.44
Formula Weight 354.44
Percent Purity 99.68%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.