missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Xtracta™ HMW DNA Kit

Product Code. 16406159 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
96 Purifications
Unit Size:
1 set
Product Code. No. of Reactions unitSize
16406159 96 Purifications 1 set
1 options
This item is not returnable. View return policy

Product Code. 16406159

Brand: Cytiva 29429140

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High yield extraction of high-molecular-weight genomic DNA from blood, buffy coat, saliva, cultured mammalian cells and solid tissue with a simplified protocol.

Sera-Xtracta™ HMW DNA Kit offers magnetic bead-based extraction and purification of genomic DNA from whole blood (treated with ethylenediaminetetraacetic acid (EDTA), citrate or heparin), buffy coat, saliva, cultured mammalian cells and solid tissue samples. The extraction protocols are designed to efficiently remove contaminants and PCR inhibitors while minimizing shearing, resulting in high quality high-molecular-weight genomic DNA with minimal RNA carryover.

  • Suitable for long-read sequencing: Isolates high-molecular-weight DNA > 200 kb.
  • Efficient scale-up and flexibility: Provides robust chemistry for sample input volumes from 200 μL – 2 mL.
  • Efficient removal of inhibitors to give high purity genomic DNA: Typical purity ratio (A260/A280 and A260/A230) of > 1.7.
  • Automation friendly reagents and easy to use protocols
  • Simplified protocol minimizes the co-purification of RNA.
  • Compatible with molecular biology techniques, including next-generation sequencing (NGS), cloning, restriction enzyme digestion, PCR amplification and genotyping applications.

The kit uses chaotropic agents to extract DNA from samples, denature protein components and promote the selective binding of DNA to the silica-coated magnetic beads. Proteinase K is the protease of choice to digest protein from samples, because it is active even when enzyme inhibitors such as EDTA and detergents are present in samples. Denatured contaminants are easily removed by subsequent washing of the silica beads with an ethanolic buffer set. The purified genomic DNA is eluted in a low ionic strength buffer at a concentration suitable for most downstream molecular biology applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Proteinase K 60 mg (dried); Lysis buffer 22 mL; Silica-coated magnetic beads 1.5 mL; Binding buffer 26 mL; Wash 1 buffer AQ 120 mL; Wash buffer 2 AQ 30mL; gDNA Elution buffer 14mL. Store at room temperature.
Description Approx. 12 μg binding capacity based on specified 15 μL aliquot of supplied magnetic bead suspension = 1.5 mg beads at 100 mg/mL
Elution Volume 100 μL
Includes Proteinase K 3mL, Lysis buffer 22mL, Silica-coated magnetic beads: 1.6mL, Binding buffer 26mL, Wash buffer 1 AQ 120mL, Wash buffer 2 AQ 30mL, gDNA Elution buffer 14mL
No. of Reactions 96 Purifications
Purity Purity ratio (A260/A280) > 1.7
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sera-Xtracta Genomic DNA Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : guanidinium chloride, trometamol, proteases
  • UNKNOWN ACUTE-70 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 70% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.