missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag™ Streptavidin Magnetic Beads, high biotin binding capacity
Shop All Cytiva Products
Click to view available options
Quantity:
1 mL
5 mL
100 mL
Unit Size:
Each
Description
Biotin Binding Capacity of 4500 to 5500pmol/mg beads
Specifications
Specifications
| Concentration | 1% solids (10mg/mL) |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets |
| Quantity | 1 mL |
| Diameter (Metric) | 1 μm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction