missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Select

Delivers selective binding of DNA fragments for use with next-generation sequencing (NGS) and PCR clean-up.

Brand:  Cytiva 29343057

Additional Details : Weight : 0.65450kg

 View more versions of this product

Product Code. 16427655

  • £3112.00 / 450mL
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sera-Mag Select PCR clean-up and size selection reagent combines the exceptional binding characteristics of Sera-Mag Carboxyl Speedbeads with an optimized binding solution in a ready-to-use formulation. The DNA size range isolated from the process can be tailored to suit the user’s end requirements by adjusting the amount of reagent that is mixed with a fixed volume of sample.

  • Use a single reagent for PCR clean-up and size selection
  • Easily control what size DNA is retained or eliminated by adding various ratios of Sera-Mag Select to your sample, enabling NGS library and PCR fragment clean-up, as well as dual size selection of libraries
  • Uses the same protocols as existing reagents so it can be directly integrated into existing workflows with minimal disruption
  • Sera-Mag Select delivers tight distribution in size selection for high yield recovery of specific fragment sizes
  • Suitable for manual, high throughput and automated workflows
  • Sera-Mag Select should be stored in the refrigerator between 2°C to 8°C - Do not freeze - Store upright and keep bottle tightly sealed
Specifications

Specifications

2°C to 8°C
Tested functionally in size selection
450 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers