missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophobic

Product Code. 11879782 Shop All Cytiva Products
Click to view available options
Quantity:
15 mL
100 mL
1 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11879782

Brand: Cytiva 44152105050350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.

  • High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophobic bead based on crosslinking requirements for the encapsulation layer
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5% solids (50mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) DNA Sample Prep, Protein Purification
Quantity 100 mL
Diameter (Metric) 1 μm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.