missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SeqStudio™ Integrated Capillary Protector

Product Code. 15899876 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15899876 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15899876 Supplier Applied Biosystems™ Supplier No. A31923
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

The Applied Biosystems SeqStudio Integrated Capillary Protector (ICP) is used on the SeqStudio Cartridge. Gel-filled and is for one-time-use only.

The Applied Biosystems™ SeqStudio™ Integrated Capillary Protector (ICP) is used on the SeqStudio Cartridge. The Integrated Capillary Protector protects the capillary ends on the cartridge. The protector is gel-filled and is for one-time-use only.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Product Line SeqStudio
Type Integrated Capillary Protector
Quantity 1 each
Shipping Condition Approved for shipment at room temperature or wet ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.