missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SeqStudio™ Genetic Analyzer System with SmartStart

Product Code. p-7138028
Change view
Click to view available options
Content And Storage:
System + SmartStart + 1 yr ext warranty
System + SmartStart + 3 yr ext warranty
Unit Size:
1 unit
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage unitSize
15925322 System + SmartStart + 1 yr ext warranty 1 unit
16834580 System + SmartStart + 3 yr ext warranty Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15925322 Supplier Applied Biosystems™ Supplier No. A35645
Add to basket
Add to basket
This item is not returnable. View return policy

The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis.

  • The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis
  • Combining an extended warranty with your system purchase, Help extend the life of your investment. These packages include the system, SmartStart, and an extended warranty.
  • Simplified instrument software with interactive touch screen for greater ease of use
  • Minimized hands-on setup time with integrated reagents cartridge (SeqStudio Cartridge)
  • Application flexibility to run Sanger sequencing and fragment analysis on the same run
  • Connectivity: Wi-Fi or wired Ethernet connection
  • Web-enabled remote monitoring capability
  • Consumables usage tracking through radio frequency identification (RFID)
  • SmartStart is performed by a Field Application Scientist (FAS) in your lab. SmartStart helps you:
  • Get up and running quickly
  • Get connected to your local network and Cloud
  • Choose appropriate reagents and consumables to meet your needs
  • Content will vary depending on your level of knowledge and experience.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage System + SmartStart + 1 yr ext warranty
Product Line SeqStudio
Type Genetic Analyzer
Duration 1 Year Extended Warranty
Shipping Condition Room Temperature
Have you tested the SeqStudio Genetic Analyzer (Cat. No. A35644, A35645) in low humidity conditions (below 20%)?

SeqStudio Genetic Analyzer has not been tested for operation in low humidity conditions and is validated for environments with 20-80% humidity. Therefore, we do not recommend using SeqStudio Genetic Analyzer in below 20% humidity.

If the computer attached to the SeqStudio Genetic Analyzer System with SmartStart is upgraded to Win 10, does the data collection software need to be upgraded as well?

No, since the SeqStudio instrument's data collection software is on the instrument itself, it is not necessary that you upgrade it.

Can I run SNaPshot applications, between 35 and 120 bp, on the SeqStudio Genetic Analyzer and the Applied Biosystems 3500 Genetic Analyzer with a 50 cm capillary array?

Both the SeqStudio Genetic Analyzer and the Applied Biosystems 3500 Genetic Analyzer have run modules specific for the SNaPshot application.

- For the 3500 Genetic Analyzer, the run module is "SNaPshot50_POP7" and it has a ~30 minute run time.
- For the SeqStudio Genetic Analyzer, the run module is "SNaPshot", the optimal size range is 40-120 bp, and it runs for ~25 minutes but the 35 bp fragment will be collected.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.