missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SeqStudio™ Genetic Analyzer System with SmartStart

Product Code. 16892250 Shop All Applied Biosystems Products
Change view
Click to view available options
Content And Storage:
System + SmartStart
System + SmartStart + 1 yr ext warranty
System + SmartStart + 3 yr ext warranty
Unit Size:
1 unit
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage unitSize
16892250 System + SmartStart Each
15925322 System + SmartStart + 1 yr ext warranty 1 unit
16834580 System + SmartStart + 3 yr ext warranty Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16892250 Supplier Applied Biosystems™ Supplier No. undefined

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis.

  • The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis
  • Combining an extended warranty with your system purchase, Help extend the life of your investment. These packages include the system, SmartStart, and an extended warranty.
  • Simplified instrument software with interactive touch screen for greater ease of use
  • Minimized hands-on setup time with integrated reagents cartridge (SeqStudio Cartridge)
  • Application flexibility to run Sanger sequencing and fragment analysis on the same run
  • Connectivity: Wi-Fi or wired Ethernet connection
  • Web-enabled remote monitoring capability
  • Consumables usage tracking through radio frequency identification (RFID)
  • SmartStart is performed by a Field Application Scientist (FAS) in your lab. SmartStart helps you:
  • Get up and running quickly
  • Get connected to your local network and Cloud
  • Choose appropriate reagents and consumables to meet your needs
  • Content will vary depending on your level of knowledge and experience.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage System + SmartStart
Product Line SeqStudio
Type Genetic Analyzer
Duration 1-day training
Format 96-well Plate, 8-tube Strip
Shipping Condition Room Temperature
Software Type Fragment Analysis
Length (English) 19.48 in.
Length (Metric) 49.5 cm
Width (English) 25.5 in.
Width (Metric) 64.8 cm
Height (English) 17.4 in.
Height (Metric) 44.2 cm
Electrical Requirements 100 V to 240 V
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.