Learn More
Applied Biosystems™ SeqStudio™ Genetic Analyzer System with SmartStart
Description
- The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis
- Combining an extended warranty with your system purchase, Help extend the life of your investment. These packages include the system, SmartStart, and an extended warranty.
- Simplified instrument software with interactive touch screen for greater ease of use
- Minimized hands-on setup time with integrated reagents cartridge (SeqStudio Cartridge)
- Application flexibility to run Sanger sequencing and fragment analysis on the same run
- Connectivity: Wi-Fi or wired Ethernet connection
- Web-enabled remote monitoring capability
- Consumables usage tracking through radio frequency identification (RFID)
- SmartStart is performed by a Field Application Scientist (FAS) in your lab. SmartStart helps you:
- Get up and running quickly
- Get connected to your local network and Cloud
- Choose appropriate reagents and consumables to meet your needs
- Content will vary depending on your level of knowledge and experience.
Specifications
Specifications
| Content And Storage | System + SmartStart + 1 yr ext warranty |
| Product Line | SeqStudio |
| Type | Genetic Analyzer |
| Duration | 1 Year Extended Warranty |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
SeqStudio Genetic Analyzer has not been tested for operation in low humidity conditions and is validated for environments with 20-80% humidity. Therefore, we do not recommend using SeqStudio Genetic Analyzer in below 20% humidity.
No, since the SeqStudio instrument's data collection software is on the instrument itself, it is not necessary that you upgrade it.
Both the SeqStudio Genetic Analyzer and the Applied Biosystems 3500 Genetic Analyzer have run modules specific for the SNaPshot application.
- For the 3500 Genetic Analyzer, the run module is "SNaPshot50_POP7" and it has a ~30 minute run time.
- For the SeqStudio Genetic Analyzer, the run module is "SNaPshot", the optimal size range is 40-120 bp, and it runs for ~25 minutes but the 35 bp fragment will be collected.
You may use the export and import functions to transfer settings from one installation of the SAE Administrator Console to another.
Archiving audit records removes the records from the SAE Administrator Console and saves them in an internally specified location on the same computer on which the SAE Administrator Console is installed. Note: Archived audit records are accessible for viewing in the SAE Administrator Console.
For Research Use Only. Not for use in diagnostic procedures.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
