missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SeqStudio™ Cathode Buffer Container

Product Code. 15809886 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 containers
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15809886 4 containers Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15809886 Supplier Applied Biosystems™ Supplier No. A33401
  • ONLINE EXCLUSIVE PRICE
    £148.00
    £133.65 / Pack of 4
    Save £14.35
    9% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems SeqStudio Cathode Buffer Container (CBC) contains running buffer to support Sanger sequencing and fragment analysis applications on the SeqStudio Genetic Analyzer System.

  • Ready-to-use, disposable container with running buffer
  • Buffer usage tracking with radio frequency identification (RFID)
  • Two week on-instrument shelf life after installation

Order Info

4 containers

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Product Line SeqStudio
Type Cathode Buffer Container
Quantity 4 containers
Shipping Condition Approved for shipment on Wet or Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.