missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SeqStudio™ Cartridge

Product Code. 15819886 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
125 injections/500 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15819886 125 injections/500 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819886 Supplier Applied Biosystems™ Supplier No. A33671
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems SeqStudio Cartridge is an all-in-one and easy-to-use reagent cartridge that includes capillaries, universal polymer, buffer, and pump required to run on the SeqStudio Genetic Analyzer System.

  • Minimize hands-on setup time with all-in-one cartridge
  • Track consumables usage through radio frequency identification (RFID)
  • Allows for 125 injections or 500 rxns per cartridge
  • Responsible, simple, no-charge recycling program

Order Info

125 injections/500 rxns

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Capillary Length 28 cm
For Use With (Application) Rare Variant Sequencing, Resequencing, Comparative Sequencing (Somatic Mutations - 10:50), Fragment Analysis (Denaturing), Short Read Sequencing, De Novo Sequencing - Low to Mid Throughput, Short PCR Product Sequencing
Product Line SeqStudio
Type Reagent Cartridge
Quantity 125 injections/500 reactions
Shipping Condition Wet Ice
How do I recycle my SeqStudio cartridge if my country is not supported by the recycling program?

We would recommend disposing of your SeqStudio cartridge according to your country's regulations on proper disposal of laboratory equipment and reagents.

How do I know if the SeqStudio cartridge recycling program is supported in my country?

You can view the details and availability of the SeqStudio Cartridge recycling program on this web page (https://www.thermofisher.com/us/en/home/life-science/sequencing/sanger-sequencing/sanger-sequencing-technology-accessories/seqstudio-genetic-analyzer/seqstudio-cartridge-recycling-program.html) or speak to your local representative for more information.

Where does the recycling of my used SeqStudio cartridge take place?

We have partnered with E-End Solutions in North America and SIMMs Recycling Solutions in Europe for recycling of used SeqStudio cartridges. More details about the program can be found in the SeqStudio Cartridge Green Fact Sheet (https://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/seqstudio-cartridge-green-fact-sheet.pdf).

What should I do if I lost or misplaced my SeqStudio cartridge recycling return envelope?

If the original SeqStudio cartridge recycling return envelope is not available, you can place the return label on another polyethylene envelope of similar dimensions. The envelope dimensions are 12" x 15 1⁄2".

What is the cost to recycle my SeqStudio cartridge?

There is no cost to recycle your SeqStudio cartridge when you use the pre-paid label provided by Thermo Fisher Scientific.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.