Learn More
Applied Biosystems™ SeqScape™ Software v4.1, initial license
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 license
Unit Size:
Each
Description
- SeqScape Software is a resequencing package designed for mutation detection and analysis, SNP discovery and validation, pathogen sub-typing, allele identification, and sequence confirmation
- It provides library functions for comparison to a known group of sequences, as well as 21 CFR Part 11 functionalities
Specifications
Specifications
| Product Line | SeqScape |
| Software Type | Sequencing Data Analysis |
| Content And Storage | The software is provided in the form of a USB stick. |
| Data Entry | CE Sequencing (.ab1 files) |
| License | Initial |
| Operating System | Windows 10 |
| Product Type | SeqScape Software v4.0 Initial License |
| Quantity | 1 license |
| Software Category | Supports files from all AB Genetic Analyzers |
For Research Use Only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto