missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Septum for 10-425 screw thread cap

Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.26200kg

 View more versions of this product

Product Code. 11834820

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Septa
Specifications

Specifications

Seota
9.5 mm
1000
Red, White
PTFE, Silicone
1 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers