missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Septum for 10-425 screw thread cap

Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 6.01000kg

 View more versions of this product

Product Code. 10422614

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Septa
Specifications

Specifications

Septa
10 mm
1000
White
PTFE Virginal
0.25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers