missing translation for 'onlineSavingsMsg'
Learn More

Septin 9 Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

£119.00 - £295.00

Specifications

Antigen Septin 9
Clone 1C6F10
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234775
View Documents
Proteintech
67548-1-IG-20UL
20 μL
ONLINE EXCLUSIVE PRICE
£135.00
£119.00 / 20µL
Save £16.00
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30234616
View Documents
Proteintech
67548-1-IG-150UL
150 μL
ONLINE EXCLUSIVE PRICE
£349.00
£295.00 / 150µL
Save £54.00
15% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Septin 9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Septin 9. This antibody recognizes human, pig, rat antigen. The Septin 9 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

Septin 9
1000 μg/mL
Monoclonal
Liquid
RUO
10801
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
1C6F10
Western Blot, ELISA
Unconjugated
Mouse
Human, Pig, Rat
AF17q25; FLJ75490; KIAA0991; MSF; MSF1; NAPB; PNUTL4; SINT1; SeptD1
IgG1
Protein G purification
Monoclonal Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.