missing translation for 'onlineSavingsMsg'
Learn More

Septin-9 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18408606 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408606 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408606 Supplier Novus Biologicals Supplier No. NBP128711FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Septin-9 Polyclonal antibody specifically detects Septin-9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-9
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AF17q25, FLJ75490, KIAA0991Ov/Br septin, MLL septin-like fusion protein, MLL septin-like fusion protein MSF-A, MSF1NAPB, MSFMLL septin-like fusion, Ovarian/Breast septin, PNUTL4, SeptD1, septin 9, Septin D1, septin-9, SINT1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 536 and 586 of human septin 9 using the numbering given in entry NP_001106963.1 (GeneID 10801).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10801
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.