missing translation for 'onlineSavingsMsg'
Learn More

Septin-9 Antibody (2C6), Novus Biologicals™

Product Code. 18399439 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399439 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399439 Supplier Novus Biologicals Supplier No. H00010801M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Septin-9 Monoclonal antibody specifically detects Septin-9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-9
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2C6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100 to 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 to 1:500, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH21192
Gene Alias AF17q25, FLJ75490, KIAA0991Ov/Br septin, MLL septin-like fusion protein, MLL septin-like fusion protein MSF-A, MSF1NAPB, MSFMLL septin-like fusion, Ovarian/Breast septin, PNUTL4, SeptD1, septin 9, Septin D1, septin-9, SINT1
Host Species Mouse
Immunogen SEPT9 (AAH21192, 26 a.a. ~ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PRRVQTPLLRATVASSTQKFQDLGVKNSEPSARHVDSLSQRSPKASLRRVELSGPKAAEPVSRRTELSIDISSKQVENAGAIGPSRFGLKRAEVLGHKTP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10801
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.