missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-8 Antibody (9H10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43033
This item is not returnable.
View return policy
Description
Septin-8 Monoclonal antibody specifically detects Septin-8 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Septin-8 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| 9H10 | |
| Western Blot 1:500 - 1:2000, ELISA | |
| KIAA0202SEP2, septin 8, septin-8 | |
| septin 8 - (Uniprot# Q92599) | |
| 100 μg | |
| Stem Cell Markers | |
| 23176 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction