missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Septin-7 Recombinant Rabbit Monoclonal Antibody (K01_1Y25)
Description
Immunogen sequence: RNMLIRTHMQ DLKDVTNNVH YENYRSRKLA AVTYNGVDNN KNKGQLTKSP LAQMEEERRE HVAKMKKMEM EMEQVFEMKV KEKVQKLKDS EAELQRRHEQ MKKNLEAQHK ELEEKRRQFE DEKANWEAQQ RILEQQNSSR TLEKNKKKGK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%, Pig - 100%, Cynomolgus monkey - 100%.
This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.
Specifications
Specifications
| Antigen | Septin-7 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1Y25 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | SEPTIN7 |
| Gene Accession No. | O55131, Q16181, Q9WVC0 |
| Gene Alias | 7-Sep; Cdc10; CDC10 (cell division cycle 10, S. cerevisiae, homolog); CDC10 (cell division cycle 10, S.cerevisiae, homolog); CDC10 protein homolog; CDC3; cell division cycle 10 homolog; E430034N22; NBLA02942; SEPT7; SEPT7A; septin 7; septin 7 variant 4; Septin7; septin-7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?