missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Septin-7 Recombinant Rabbit Monoclonal Antibody (K01_1Y25)
GREENER_CHOICE

Product Code. 30825595
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825595 100 μL
30829584 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825595 Supplier Invitrogen™ Supplier No. MA563806

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: RNMLIRTHMQ DLKDVTNNVH YENYRSRKLA AVTYNGVDNN KNKGQLTKSP LAQMEEERRE HVAKMKKMEM EMEQVFEMKV KEKVQKLKDS EAELQRRHEQ MKKNLEAQHK ELEEKRRQFE DEKANWEAQQ RILEQQNSSR TLEKNKKKGK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%, Pig - 100%, Cynomolgus monkey - 100%.

This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-7
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Y25
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene SEPTIN7
Gene Accession No. O55131, Q16181, Q9WVC0
Gene Alias 7-Sep; Cdc10; CDC10 (cell division cycle 10, S. cerevisiae, homolog); CDC10 (cell division cycle 10, S.cerevisiae, homolog); CDC10 protein homolog; CDC3; cell division cycle 10 homolog; E430034N22; NBLA02942; SEPT7; SEPT7A; septin 7; septin 7 variant 4; Septin7; septin-7
Gene Symbols SEPTIN7
Host Species Rabbit
Immunogen Recombinant protein of human SEPT7 (aa 286-435).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235072, 64551, 989
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.