missing translation for 'onlineSavingsMsg'
Learn More

Septin-7 Rabbit anti-Human, Mouse, Rat, Clone: S09-6A1, Novus Biologicals™

Product Code. 18326546 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326546 25 μg 25µL
18317946 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18326546 Supplier Novus Biologicals Supplier No. NBP31509025UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Septin-7 Monoclonal antibody specifically detects Septin-7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-7
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone S09-6A1
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry-Paraffin 20 μg/mL, Immunofluorescence 20 μg/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias CDC10 cell division cycle 10 homolog, CDC10 cell division cycle 10 homolog (S. cerevisiae), CDC10 protein homolog, CDC10S. cerevisiae, homolog), Nbla02942, septin 7, septin-7
Host Species Rabbit
Immunogen Recombinant protein of human Septin-7 (Uniprot#: Q16181)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 989
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.