missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-7 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-06077
This item is not returnable.
View return policy
Description
Septin-7 Polyclonal specifically detects Septin-7 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Septin-7 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CDC10 cell division cycle 10 homolog, CDC10 cell division cycle 10 homolog (S. cerevisiae), CDC10 protein homolog, CDC10S. cerevisiae, homolog), Nbla02942, septin 7, septin-7 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize all three reported isoforms (NP_001779.3; NP_001011553.2; NP_001229885.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_001779.3 | |
| SEPTIN7 | |
| Peptide with sequence C-DNNKNKGQLTKSP corresponding to internal region according to NP_001011553.2, NP_001229885.1. | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 989 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction