missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-6 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-51930
This item is not returnable.
View return policy
Description
Septin-6 Polyclonal specifically detects Septin-6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| Septin-6 | |
| Polyclonal | |
| Western Blot 0.1 - 1 μg/mL, Immunohistochemistry 3 - 5 μg/mL, Immunohistochemistry-Paraffin 3 - 5 μg/mL, Peptide ELISA Detection limit 1:4000 | |
| NP_665798.1 | |
| SEPTIN6 | |
| Peptide with sequence C-DEVNAFKQRKTA corresponding to internal region according to NP_665798.1, NP_055944.2, NP_665801.1. | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 23157 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| KIAA0128SEP2septin 2, MGC16619, MGC20339, SEPT2, septin 6, septin-6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize the reported isoforms NP_665798.1, NP_055944.2 and NP_665801.1. Reported variants NP_665799.1 and NP_665798.1 represent identical protein. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction