missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-5 Antibody (SP18), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-81058FR
This item is not returnable.
View return policy
Description
Septin-5 Monoclonal specifically detects Septin-5 in Human, Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Septin-5 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| SEPTIN5 | |
| Immunoprecipitate from human brain. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 5413 | |
| Human, Rat, Rabbit | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| SP18 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CDCREL, CDCREL1, CDCREL-1, cell division control related protein 1, Cell division control-related protein 1, HCDCREL-1, peanut-like 1, peanut-like 1 (Drosophila), Peanut-like protein 1, platelet glycoprotein Ib beta chain, PNUTL1H5, septin 5, septin-5 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody recognises Septin-5 (CDCrel-1) and does not cross-react with CDCrel-2. | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction