missing translation for 'onlineSavingsMsg'
Learn More

Septin-5 Antibody (SP18), Biotin, Novus Biologicals™

Product Code. 18340851 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340851 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340851 Supplier Novus Biologicals Supplier No. NBP281058B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Septin-5 Monoclonal specifically detects Septin-5 in Human, Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-5
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone SP18
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS
Gene Alias CDCREL, CDCREL1, CDCREL-1, cell division control related protein 1, Cell division control-related protein 1, HCDCREL-1, peanut-like 1, peanut-like 1 (Drosophila), Peanut-like protein 1, platelet glycoprotein Ib beta chain, PNUTL1H5, septin 5, septin-5
Gene Symbols SEPTIN5
Host Species Mouse
Immunogen Immunoprecipitate from human brain.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5413
Test Specificity This antibody recognises Septin-5 (CDCrel-1) and does not cross-react with CDCrel-2.
Target Species Human, Rat, Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.