missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00 - £398.00
Specifications
| Antigen | Septin-4 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/ Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18583263
|
Novus Biologicals
NBP1-76667 |
0.1 mg |
£398.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18507162
|
Novus Biologicals
NBP1-76667-0.025mg |
0.025 mg |
£134.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Septin-4 Polyclonal antibody specifically detects Septin-4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Septin-4 | |
| Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/ Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AAG45673 | |
| 5414 | |
| IgG | |
| Peptide affinity purified |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS | |
| Apoptosis-related protein in the TGF-beta signaling pathway, ARTSPeanut-like protein 2, BRADEION, Bradeion beta, Brain protein H5, CE5B3, Cell division control-related protein 2, Cerebral protein 7, H5, hCDCREL-2SEP4, hucep-7, MART, peanut-like 2, peanut-like 2 (Drosophila), PNUTL2CE5B3 beta, septin 4, septin-4, septin-M | |
| Antibody was raised against a synthetic peptide corresponding to amino acids near the amino terminus of human ARTS. The immunogen is located within the first 50 amino acids of ARTS. Amino Acid Squence: KRFLEDTTDDGE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title