missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Septin 3 Monoclonal Antibody (OTI3H6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527418
This item is not returnable.
View return policy
Description
Septin 3 Monoclonal Antibody for Western Blot, IHC (P)
This gene belongs to the septin family of GTPases. Members of this family are required for cytokinesis. Expression is upregulated by retinoic acid in a human teratocarcinoma cell line. The specific function of this gene has not been determined. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Specifications
| Septin 3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UH03, Q9Z1S5 | |
| SEPT3 | |
| Full length human recombinant protein of SEPT3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H6 | |
| Unconjugated | |
| SEPT3 | |
| 3110018K01Rik; 3-Sep; AV154067; B530002E20Rik; bK250D10.3; CTA-250D10.3; G-septin; G-septin alpha; MGC133218; neuronal-specific septin 3; neuronal-specific septin-3; OTTHUMP00000028742; P40; Sep3; SEPT3; septin 3; Septin3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 24050, 55964 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction