missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-2 Rabbit anti-Human, Mouse, Rat, Clone: 9N1B3, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Septin-2 |
|---|---|
| Clone | 9N1B3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18383754
|
Novus Biologicals
NBP3-16653-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18359594
|
Novus Biologicals
NBP3-16653-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Septin-2 Monoclonal antibody specifically detects Septin-2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Septin-2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4735 | |
| IgG | |
| Affinity purified |
| 9N1B3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DIFF6NEDD-5, hNedd5, KIAA0158Neural precursor cell expressed developmentally down-regulated protein 5, NEDD5Pnutl3, neural precursor cell expressed, developmentally down-regulated 5, septin 2, septin-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 202-298 of human Septin-2 (Q15019). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title