missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Septin-14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567282
This item is not returnable.
View return policy
Description
Immunogen sequence: QLPTDEETAAQ ANSSVSGLL Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 55%.
SEPT14 is a member of the highly conserved septin family of GTP-binding cytoskeletal proteins implicated in membrane transport, apoptosis, cell polarity, cell cycle regulation, cytokinesis, and other cellular functions.[supplied by OMIM, Jul 2008].
Specifications
| Septin-14 | |
| Polyclonal | |
| Unconjugated | |
| SEPT14 | |
| 14-Sep; 1700016K13Rik; Sept14; septin 14; SEPTIN14; septin-14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 346288 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZU15 | |
| SEPT14 | |
| Recombinant Human Septin-14. Recombinant protein control fragment (Product #RP-107920). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction