missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | Septin-11 |
|---|---|
| Dilution | Western Blot 1:2000 - 1:6000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30226945
|
Novus Biologicals
NBP3-35205-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230640
|
Novus Biologicals
NBP3-35205-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Septin-11 Polyclonal antibody specifically detects Septin-11 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Septin-11 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 55752 | |
| IgG | |
| Affinity purified |
| Western Blot 1:2000 - 1:6000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ10849, septin 11, septin-11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human Septin-11 (NP_060713.1).,, Sequence:, MAVAVGRPSNEELRNLSLSGHVGFDSLPDQLVNKSTSQGFCFNILCVGETGIGKSTLMDTLFNTKFESDPATHNEPGVRLKARSYELQESNVRLKLTIVDTVGFGDQINKDDSYKPIVEY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title